Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ROPN1
Entrez Gene ID
100359353
Gene Name
rhophilin associated tail protein 1
Gene Aliases
ROPN1B
Chromosome Location
Chr.13: 136395628 - 136428170 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172369.1 NP_001165840.1 4 - 5 604 74
XM_021068368.1 XP_020924027.1 - 628 74
XM_021068369.1 XP_020924028.1 - 693 74
GenBank mRNA AK240561.1 - 4 - 5 599 74
GU373671.1 - 4 - 5 604 74

View Full Product Details