Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HES1
Entrez Gene ID
100499567
Gene Name
hes family bHLH transcription factor 1
Gene Aliases
-
Chromosome Location
Chr.13: 131135970 - 131137865 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195231.1 NP_001182160.1 2 - 3 213 76
GenBank mRNA AK233503.1 - 2 - 3 449 76
AK238852.1 - 2 - 3 457 76
AK389479.1 - 2 - 3 456 76
AK395300.1 - 2 - 3 447 76
HM235721.1 - 2 - 3 213 76

View Full Product Details