Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BMP3
Entrez Gene ID
100512974
Gene Name
bone morphogenetic protein 3
Gene Aliases
-
Chromosome Location
Chr.8: 136880456 - 136904739 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206388.1 NP_001193317.1 1 - 2 531 71
GenBank mRNA HQ127224.1 - 1 - 2 531 71

View Full Product Details