Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJC5B
Entrez Gene ID
100157247
Gene Name
DnaJ heat shock protein family (Hsp40) member C5 beta
Gene Aliases
CSP-beta
Chromosome Location
Chr.4: 68557670 - 68641134 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206401.1 NP_001193330.1 1 - 2 105 76
GenBank mRNA CV874035.1 - 1 - 2 79 76

View Full Product Details