Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPATCH4
Entrez Gene ID
100152732
Gene Name
G-patch domain containing 4
Gene Aliases
-
Chromosome Location
Chr.4: 93486365 - 93492837 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243353.1 NP_001230282.1 2 - 3 210 95
GenBank mRNA AK344967.1 - 2 - 3 210 95
AK351608.1 - 2 - 3 209 95
AK400238.1 - 2 - 3 196 95
AK400822.1 - 2 - 3 210 95
FS641076.1 - 2 - 3 210 95
FS700194.1 - 2 - 3 207 95

View Full Product Details