Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPATCH4
Entrez Gene ID
100152732
Gene Name
G-patch domain containing 4
Gene Aliases
-
Chromosome Location
Chr.4: 93486365 - 93492837 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243353.1 NP_001230282.1 5 - 6 470 65
GenBank mRNA AK344967.1 - 5 - 6 470 65
AK351608.1 - 5 - 6 469 65
AK400238.1 - 5 - 6 456 65
AK400822.1 - 5 - 6 470 65
FS700194.1 - 5 - 6 467 65

View Full Product Details