Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C5H12orf4
Entrez Gene ID
100153309
Gene Name
chromosome 5 C12orf4 homolog
Gene Aliases
-
Chromosome Location
Chr.5: 65896750 - 65944183 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243412.1 NP_001230341.1 3 - 4 407 107
NM_001243413.1 NP_001230342.1 2 - 3 264 107
XM_005655700.3 XP_005655757.1 - 1047 107
XM_013988430.2 XP_013843884.1 - 1132 107
XM_013988431.2 XP_013843885.1 - 306 107
XM_021090919.1 XP_020946578.1 - 1053 107
XM_021090921.1 XP_020946580.1 - 296 107
XM_021090922.1 XP_020946581.1 - 1132 107
GenBank mRNA AK343836.1 - 3 - 4 391 107
AK400126.1 - 3 - 4 390 107
EB414072.1 - 2 - 3 236 107
FS706643.1 - 3 - 4 407 107

View Full Product Details