Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GNB1L
Entrez Gene ID
100153889
Gene Name
G protein subunit beta 1 like
Gene Aliases
-
Chromosome Location
Chr.14: 51306561 - 51343040 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243455.1 NP_001230384.1 3 - 4 560 105
XM_005670934.3 XP_005670991.1 - 571 105
XM_021071921.1 XP_020927580.1 - 379 105
GenBank mRNA BP158014.1 - 3 - 4 560 105
CJ029038.1 - 3 - 4 415 105

View Full Product Details