Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPL15
Entrez Gene ID
100155047
Gene Name
mitochondrial ribosomal protein L15
Gene Aliases
-
Chromosome Location
Chr.4: 77069074 - 77083595 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243528.1 NP_001230457.1 2 - 3 548 100
XM_021088564.1 XP_020944223.1 - 1029 100
GenBank mRNA AK236429.1 - 2 - 3 284 100
AK240283.1 - 2 - 3 320 100
AK398677.1 - 3 - 4 929 100
AK400153.1 - 3 - 4 767 100

View Full Product Details