Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXTL2
Entrez Gene ID
100155305
Gene Name
exostosin like glycosyltransferase 2
Gene Aliases
-
Chromosome Location
Chr.4: 117396601 - 117420707 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243549.1 NP_001230478.1 1 - 2 568 130
XM_013988168.2 XP_013843622.1 - 961 130
GenBank mRNA AK343419.1 - 1 - 2 568 130
DN118116.1 - 1 - 2 164 130

View Full Product Details