Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SULT2B1
Entrez Gene ID
100626471
Gene Name
sulfotransferase family 2B member 1
Gene Aliases
-
Chromosome Location
Chr.6: 53928248 - 53963443 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243697.1 NP_001230626.1 2 - 3 319 92
GenBank mRNA AK349606.1 - 2 - 3 319 92

View Full Product Details