Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXOSC7
Entrez Gene ID
100511347
Gene Name
exosome component 7
Gene Aliases
-
Chromosome Location
Chr.13: 28176806 - 28206435 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243824.1 NP_001230753.1 4 - 5 431 77
GenBank mRNA AK395369.1 - 4 - 5 450 77
AY610143.1 - 4 - 5 431 77

View Full Product Details