Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD38
Entrez Gene ID
100511702
Gene Name
CD38 molecule
Gene Aliases
-
Chromosome Location
Chr.8: 11083682 - 11130783 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243883.1 NP_001230812.1 5 - 6 961 71
GenBank mRNA AJ236934.1 - 5 - 6 921 71
AK231339.1 - 5 - 6 961 71
AK237000.1 - 5 - 6 927 71

View Full Product Details