Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM32A
Entrez Gene ID
100511743
Gene Name
family with sequence similarity 32 member A
Gene Aliases
-
Chromosome Location
Chr.2: 61388628 - 61394251 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243887.1 NP_001230816.1 3 - 4 298 88
GenBank mRNA AK239819.1 - 3 - 4 298 88
AK395834.1 - 3 - 4 310 88
AK400888.1 - 3 - 4 298 88

View Full Product Details