Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MARVELD2
Entrez Gene ID
100516404
Gene Name
MARVEL domain containing 2
Gene Aliases
-
Chromosome Location
Chr.16: 47529857 - 47563485 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243948.1 NP_001230877.1 1 - 2 157 72
GenBank mRNA AK396289.1 - 1 - 2 132 72
FS700099.1 - 1 - 2 157 72

View Full Product Details