Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC42
Entrez Gene ID
100514007
Gene Name
coiled-coil domain containing 42
Gene Aliases
-
Chromosome Location
Chr.12: 53869559 - 53883929 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244077.1 NP_001231006.1 1 - 2 90 88

View Full Product Details