Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GNPDA1
Entrez Gene ID
100512094
Gene Name
glucosamine-6-phosphate deaminase 1
Gene Aliases
-
Chromosome Location
Chr.2: 143604362 - 143616037 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244093.1 NP_001231022.1 4 - 5 459 92
XM_013995256.2 XP_013850710.1 - 990 92
GenBank mRNA AK349993.1 - 4 - 5 450 92

View Full Product Details