Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MXD3
Entrez Gene ID
100512157
Gene Name
MAX dimerization protein 3
Gene Aliases
-
Chromosome Location
Chr.2: 80636778 - 80641346 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244104.1 NP_001231033.1 2 - 3 578 110
XM_005661448.1 XP_005661505.1 - 783 110
GenBank mRNA AK232358.1 - 2 - 3 240 110
AK234563.1 - 2 - 3 577 110
AK235641.1 - 2 - 3 244 110
BW970534.1 - 2 - 3 231 110

View Full Product Details