Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRI
Entrez Gene ID
100514123
Gene Name
sorcin
Gene Aliases
-
Chromosome Location
Chr.9: 68492235 - 68515247 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244142.1 NP_001231071.1 1 - 2 79 73
GenBank mRNA AK231770.1 - 1 - 2 67 73
AK345103.1 - 1 - 2 63 73
AK392529.1 - 1 - 2 64 73
AK395791.1 - 1 - 2 63 73
AK398866.1 - 1 - 2 63 73
AK399411.1 - 1 - 2 65 73
AK400616.1 - 1 - 2 63 73
AK400684.1 - 1 - 2 111 73
EW520500.2 - 1 - 2 111 73

View Full Product Details