Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRI
Entrez Gene ID
100514123
Gene Name
sorcin
Gene Aliases
-
Chromosome Location
Chr.9: 68492235 - 68515247 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244142.1 NP_001231071.1 6 - 7 538 66
NM_001244143.1 NP_001231072.1 6 - 7 530 66
GenBank mRNA AK231770.1 - 6 - 7 526 66
AK345103.1 - 6 - 7 522 66
AK392529.1 - 6 - 7 523 66
AK395791.1 - 6 - 7 522 66
AK398866.1 - 6 - 7 522 66
AK400616.1 - 6 - 7 522 66
AK400684.1 - 6 - 7 570 66
EW289129.2 - 6 - 7 514 66
EW326025.2 - 6 - 7 566 66
EW412279.2 - - 67 66

View Full Product Details