Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHCHD3
Entrez Gene ID
414383
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 3
Gene Aliases
-
Chromosome Location
Chr.18: 16052611 - 16348998 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244176.1 NP_001231105.1 6 - 7 689 54
XM_021078525.1 XP_020934184.1 - 711 54
XM_021078526.1 XP_020934185.1 - 717 54
XM_021078527.1 XP_020934186.1 - 711 54
XM_021078528.1 XP_020934187.1 - 711 54
XM_021078529.1 XP_020934188.1 - 710 54
XM_021078530.1 XP_020934189.1 - 717 54
GenBank mRNA AK231938.1 - 6 - 7 674 54
AK234688.1 - 6 - 7 626 54
AK348653.1 - 6 - 7 628 54
AK393133.1 - 6 - 7 677 54
AK398750.1 - 6 - 7 703 54
AY610152.1 - 6 - 7 689 54

View Full Product Details