Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THUMPD1
Entrez Gene ID
100512402
Gene Name
THUMP domain containing 1
Gene Aliases
-
Chromosome Location
Chr.3: 25285682 - 25315710 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244282.1 NP_001231211.1 2 - 3 507 108
XM_005662087.3 XP_005662144.1 - 841 108
XM_013995606.2 XP_013851060.1 - 843 108
GenBank mRNA AK235689.1 - 2 - 3 495 108
AK395562.1 - 2 - 3 507 108
FS688700.1 - 2 - 3 507 108

View Full Product Details