Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PGLYRP3
Entrez Gene ID
100626786
Gene Name
peptidoglycan recognition protein 3
Gene Aliases
-
Chromosome Location
Chr.4: 96261753 - 96277521 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244361.1 NP_001231290.1 8 - 9 1334 90
GenBank mRNA AB295597.2 - 7 - 8 989 90
AK231441.1 - 5 - 6 944 90

View Full Product Details