Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NKG7
Entrez Gene ID
100513287
Gene Name
natural killer cell granule protein 7
Gene Aliases
-
Chromosome Location
Chr.6: 58496089 - 58497827 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244529.1 NP_001231458.1 1 - 2 367 97
GenBank mRNA AK231629.1 - 1 - 2 283 97
AK237343.1 - 1 - 2 282 97
AK348727.1 - 1 - 2 289 97
AK393058.1 - 1 - 2 367 97
CJ027613.1 - 1 - 2 367 97

View Full Product Details