Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRFBP1
Entrez Gene ID
100624712
Gene Name
serum response factor binding protein 1
Gene Aliases
-
Chromosome Location
Chr.2: 125507744 - 125574751 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244566.1 NP_001231495.1 4 - 5 307 108
XM_021079835.1 XP_020935494.1 - 409 108
GenBank mRNA AK234890.1 - 4 - 5 307 108
AK390530.1 - 4 - 5 403 108

View Full Product Details