Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM26
Entrez Gene ID
100157089
Gene Name
transmembrane protein 26
Gene Aliases
-
Chromosome Location
Chr.14: 64881216 - 64925449 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244582.1 NP_001231511.1 4 - 5 682 102
GenBank mRNA AK395833.1 - 4 - 5 682 102
FS692142.1 - 4 - 5 683 102

View Full Product Details