Assay Design
The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Gene Transcripts
- Gene Symbol
- ACS1
- Entrez Gene ID
-
543470
- Gene Name
- 1-aminocyclopropane-1-carboxylate synthase
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U35779.1 |
- |
- |
248 |
92 |
View Full Product Details