Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004759.4 | NP_004750.1 | 4 | 742 | |
NM_032960.3 | NP_116584.2 | 4 | 742 | ||
XM_005273353.3 | XP_005273410.1 | 4 | 850 | ||
XM_017002810.1 | XP_016858299.1 | 4 | 408 | ||
GenBank mRNA | AK307499.1 | - | 4 | 327 | |
BC036060.1 | - | 4 | 742 | 5296259 | |
BC052584.1 | - | 4 | 725 | 6267183 | |
X75346.1 | - | 4 | 520 |