Gene Transcripts

Gene Symbol
LAMTOR3
Entrez Gene ID
8649
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 3
Gene Aliases
MAP2K1IP1, MAPBP, MAPKSP1, MP1, PRO0633, Ragulator3
Chromosome Location
Chr. 4: 99878338 - 99894546 on Build GRCh38
UniGene ID
Hs.433332
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243736.1 NP_001230665.1 6 497
NM_021970.3 NP_068805.1 6 518
NR_024170.1 5 472
GenBank mRNA AF130115.1 - 5 318
AF201947.1 - 6 428
AK223311.1 - 6 409
AK311755.1 - 6 379
AL544981.3 - 6 327
BC026245.1 - 6 411 4808855
BC110402.1 - 5 338

View Full Product Details