Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SNAP-ALPHA
- Entrez Gene ID
-
100232837
- Gene Name
- alpha-soluble NSF attachment protein
- Gene Aliases
-
GSVIVT00029393001
- Chromosome Location
-
Chr.13: 1213074 - 1218748 on Build 12X
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB001375.1 |
- |
5 - 6 |
492 |
91 |
View Full Product Details