Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VVSUC12
Entrez Gene ID
100232845
Gene Name
sucrose transporter-like
Gene Aliases
-
Chromosome Location
Chr.1_NW_002238162: 1414276 - 1424354 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF021809.1 - 1 - 2 302 83

View Full Product Details