Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SIRK
Entrez Gene ID
100233105
Gene Name
inward rectifying shaker-like K+ channel
Gene Aliases
-
Chromosome Location
Chr.10_NW_002238068: 71919 - 75858 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF359521.1 - 4 - 5 613 81

View Full Product Details