Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GS
Entrez Gene ID
100249247
Gene Name
glutamine synthetase
Gene Aliases
GSVIVT00030210001
Chromosome Location
Chr.1: 1868220 - 1872672 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AJ318053.1 - 5 - 6 402 93

View Full Product Details