Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPPS
Entrez Gene ID
100232946
Gene Name
geranyl diphosphate synthase
Gene Aliases
-
Chromosome Location
Chr.6: 16993119 - 17012872 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY351862.1 - 4 - 5 342 85

View Full Product Details