Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAP3K
Entrez Gene ID
100232947
Gene Name
mitogen-activated protein kinase kinase kinase
Gene Aliases
GSVIVT01017915001
Chromosome Location
Chr.5: 4510826 - 4524541 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY461710.1 - 4 - 5 520 104

View Full Product Details