Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CWINV
Entrez Gene ID
100232951
Gene Name
cell wall apoplastic invertase
Gene Aliases
-
Chromosome Location
Chr.9: 2103328 - 2107257 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY538262.1 - 6 - 7 1555 71

View Full Product Details