Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MADS9
Entrez Gene ID
100232978
Gene Name
PISTILLATA-like MADS-box protein-like
Gene Aliases
PI
Chromosome Location
Chr.18: 2253893 - 2256111 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA DQ059750.1 - 5 - 6 485 88

View Full Product Details