Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MADS8
Entrez Gene ID
100232995
Gene Name
MADS-box protein
Gene Aliases
GSVIVT00026312001, SOC1
Chromosome Location
Chr.15: 5947920 - 5971440 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA DQ504309.1 - 6 - 7 833 72
GU133633.1 - 6 - 7 516 72

View Full Product Details