Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GDH
Entrez Gene ID
100263479
Gene Name
L-galactose dehydrogenase
Gene Aliases
GSVIVT01036958001
Chromosome Location
Chr.3: 9110181 - 9123755 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA EF554359.1 - 2 - 3 364 84

View Full Product Details