Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DHAR
Entrez Gene ID
100233050
Gene Name
dehydroascorbate reductase
Gene Aliases
GSVIVT00029528001
Chromosome Location
Chr.13: 2255971 - 2260107 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA EU280162.1 - 2 - 3 193 80

View Full Product Details