Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYB60
Entrez Gene ID
100233072
Gene Name
R2R3 MYB60 transcription factor
Gene Aliases
-
Chromosome Location
Chr.8: 1265357 - 1266809 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA EU816358.1 - 1 - 2 134 74

View Full Product Details