Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CBL02
Entrez Gene ID
100266945
Gene Name
calcineurin B-like protein 02
Gene Aliases
GSVIVT00033435001
Chromosome Location
Chr.19: 9135621 - 9146828 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA FJ901268.1 - 3 - 4 266 101

View Full Product Details