Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CBL05
Entrez Gene ID
100249564
Gene Name
calcineurin B-like protein 05
Gene Aliases
GSVIVT00006735001
Chromosome Location
Chr.2: 1439820 - 1441257 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA FJ901271.1 - 5 - 6 375 64

View Full Product Details