Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lrrc59
Entrez Gene ID
407863
Gene Name
leucine rich repeat containing 59
Gene Aliases
-
Chromosome Location
Chr.10: 35308596 - 35313589 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001205.1 NP_001001205.1 2 - 3 207 81
XM_018097691.1 XP_017953180.1 1 - 2 888 81
GenBank mRNA BC067310.1 - 2 - 3 207 81

View Full Product Details