Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mcat
Entrez Gene ID
448008
Gene Name
malonyl-CoA-acyl carrier protein transacylase
Gene Aliases
fabd, fasn2c, mct
Chromosome Location
Chr.Un|NW_016683903: 238 - 2463 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004788.1 NP_001004788.1 - 522 83
GenBank mRNA BC074523.1 - - 522 83

View Full Product Details