Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ak2
Entrez Gene ID
448011
Gene Name
adenylate kinase 2
Gene Aliases
adk2
Chromosome Location
Chr.2: 86231354 - 86242340 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004791.1 NP_001004791.1 6 - 7 732 92
GenBank mRNA BC074526.1 - 6 - 7 732 92

View Full Product Details