Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fbxo36
Entrez Gene ID
448109
Gene Name
F-box protein 36
Gene Aliases
-
Chromosome Location
Chr.5: 125570777 - 125584637 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004838.1 NP_001004838.1 3 - 4 600 76
XM_018093829.1 XP_017949318.1 4 - 5 462 76
XM_018093830.1 XP_017949319.1 5 - 6 572 76
XM_018093831.1 XP_017949320.1 4 - 5 528 76
GenBank mRNA BC074640.1 - 3 - 4 600 76
CR761727.2 - 3 - 4 404 76

View Full Product Details