Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
tbc1d16
Entrez Gene ID
448159
Gene Name
TBC1 domain family member 16
Gene Aliases
-
Chromosome Location
Chr.10: 24770589 - 24827886 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004861.1 NP_001004861.1 5 - 6 1099 57
XM_012958813.2 XP_012814267.1 5 - 6 1090 57
XM_018097696.1 XP_017953185.1 5 - 6 1111 57
XM_018097697.1 XP_017953186.1 5 - 6 1098 57
GenBank mRNA BC074694.1 - 5 - 6 1099 57

View Full Product Details