Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hebp1
Entrez Gene ID
448253
Gene Name
heme binding protein 1
Gene Aliases
-
Chromosome Location
Chr.4: 106039391 - 106047243 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004901.1 NP_001004901.1 3 - 4 582 91
GenBank mRNA BC075317.1 - 3 - 4 582 91

View Full Product Details