Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
glrx5
Entrez Gene ID
448300
Gene Name
glutaredoxin 5
Gene Aliases
grx5
Chromosome Location
Chr.8: 67764692 - 67769016 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004919.1 NP_001004919.1 1 - 2 308 69
GenBank mRNA BC075374.1 - 1 - 2 308 69

View Full Product Details